Learn More
anti-SLC2A11, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP146401
Catalog No. NBP146401
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
SLC2A11
LYOPH
Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500
Antisera
SLC2A11
A synthetic peptide from c-terminal region of human Glucose transporter type 11 (GLUT11, SLC2A11) conjugated to an immunogenic carrier protein was used as the antigen.
Unpurified
RUO
Primary
66035
Reconstitute with 0.1 ml sterilized water to desired concentration.
Immunohistochemistry, Western Blot
Unconjugated
Whole antisera with No Preservative
facilitative glucose transporter GLUT11, glucose transporter protein 10, glucose transporter protein 11, GLUT10, GLUT-10, GLUT11, GLUT-11, SLC2A11-a, SLC2A11-c, solute carrier family 2 (facilitated glucose transporter), member 11, solute carrier family 2, facilitated glucose transporter member 11
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specific for GLUT-11.
Human