Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SLC39A4/ZIP4, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136988

Catalog No. NBP136988


Add to cart

Description

Description

SLC39A4/ZIP4 Polyclonal antibody specifically detects SLC39A4/ZIP4 in Human samples. It is validated for Peptide ELISA.
Specifications

Specifications

SLC39A4/ZIP4
0.5 mg/mL
Peptide ELISA Detection limit 1:4000
NP_060237.2
acrodermatitis enteropathica, zinc-deficiency type, FLJ20327, MGC74741, solute carrier family 39 (zinc transporter), member 4, Solute carrier family 39 member 4, zinc transporter ZIP4, ZIP-4, ZIP4AEZ, Zrt- and Irt-like protein 4
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is expected to recognize both reported isoforms (NP_060237.2; NP_570901.2)
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
SLC39A4
Peptide with sequence C-RQPKPPHEGSRAD corresponding to internal region according to NP_060237.2, NP_570901.2.
Immunogen affinity purified
RUO
Primary
55630
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only