Learn More
SLC39A6 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody rasied against synthetic peptide of SLC39A6.
Manufacturer: Abnova Corporation PAB19351
Description
Zinc is an essential cofactor for hundreds of enzymes. It is involved in protein, nucleic acid, carbohydrate, and lipid metabolism, as well as in the control of gene transcription, growth, development, and differentiation. SLC39A6 belongs to a subfamily of proteins that show structural characteristics of zinc transporters (Taylor and Nicholson, 2003 [PubMed 12659941]).[supplied by OMIM
Specifications
SLC39A6 | |
1 mg/mL | |
Rabbit polyclonal antibody rasied against synthetic peptide of SLC39A6. | |
In PBS (0.02% sodium azide) | |
Liquid | |
SLC39A6 | |
A synthetic peptide corresponding to 16 amino acids near internal region of human SLC39A6. | |
100 ug | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
Human, Murine |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Western Blot (1 ug/mL) Immunohistochemistry (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
SLC39A6 | |
LIV-1 | |
Rabbit | |
Peptide affinity purification | |
RUO | |
Primary | |
25800 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only