Learn More
MilliporeSigma™ anti-SLC3A2, Polyclonal
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma™ ST1678100UG
Catalog No. ST1678100UG
Add to cart
Description
Description
Specifically detects SLC3A2 Clone: in Human, Mouse samples, and it is validated for Immuno Blotting
This gene is a member of the solute carrier family and encodes a cell surface, transmembrane protein with anα amylase domain. The protein exists as the heavy chain of a heterodimer, covalently bound through di-sulfide bonds to one of several possible light chains. It associates with integrins and mediates integrin-dependent signaling related to normal cell growth and tumorigenesis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
Specifications
SLC3A2
Unconjugated
Purified
Full-length, human SLC3A2 (aa 1-529)
100μg
−20°C
Polyclonal
Human, Murine
Immuno Blotting, Immunoblot
In PBS, pH 7.2.
Rabbit
IgG
RUO
Primary
Recognizes the ∽50-52kDa SLC3A2 protein in mouse kidney tissue.