Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC4A7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525563

Catalog No. PIPA525563

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

SLC4A7 regulates intracellular pH and may also play a role in bicarbonate salvage in secretory epithelia. Additionally, SLC4A7 may also have associated sodium channel activity.
Specifications

Specifications

SLC4A7
Polyclonal
PBS with 0.09% sodium azide
Q9Y6M7
NBC2, NBC3, SBC2, SLC4A6, bicarbonate transporter, sodium bicarbonate cotransporter 2, sodium bicarbonate cotransporter 2b, sodium bicarbonate cotransporter 3, solute carrier family 4 member 7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9497
Immunohistochemistry, Western Blot
Unconjugated
SLC4A7
Liquid
SLC4A7
KLH conjugated synthetic peptide between 1186-1214 amino acids from the C-terminal region of human SLC4A7
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit