Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SLC7A3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178911

Catalog No. NBP178911


Add to cart

Description

Description

CAT3/SLC7A3 Polyclonal antibody specifically detects CAT3/SLC7A3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

CAT3/SLC7A3
Unconjugated
PBS (pH 7.2) and 50% Glycerol with No Preservative
Affinity Purified
SLC7A3
The rabbit was immunized with a synthetic peptide derived from human SLC7A3.
67 kDa
0.05 ml
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:1000, Flow Cytometry 1:200, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:1000
Q8WY07
ATRC3cationic amino acid transporter 3, CAT3, CAT-3Cationic amino acid transporter y+, FLJ14541, MGC20687, solute carrier family 7 (cationic amino acid transporter, y+ system), member 3, Solute carrier family 7 member 3
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
84889
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only