Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC7A9, Polyclonal, Invitrogen™

Catalog No. PIPA127907
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127907 50 μg
1 options

Catalog No. PIPA127907

Supplier: Thermo Scientific PA127907

Chicken Polyclonal Antibody

Involved in the high-affinity. sodium-independent transport of cystine and neutral and dibasic amino acids (system b(0.+)-like activity). Thought to be responsible for the high-affinity reabsorption of cystine in the kidney tubule.

PA1-27907 detects SLC7A9 from human samples.PA1-27907 may be successfully used in ELISA applications.The PA1-27715 immunogen is a synthetic peptide corresponding to human B(0.+)-type amino acid transporter 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC7A9
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Gene SLC7A9
Gene Accession No. P82251
Gene Alias BAT1, CSNU3, FLJ94301
Gene Symbols SLC7A9
Host Species Chicken
Immunogen Synthetic peptide corresponding to human B(0.+)-type amino acid transporter 1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11136
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.