Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBP50, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127670

Catalog No. PIPA127670

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Scaffold protein that connects plasma membrane proteins with members of the ezrin/moesin/radixin family and thereby helps to link them to the actin cytoskeleton and to regulate their surface expression. Necessary for recycling of internalized ADRB2. Was f

PA1-27670 detects SLC9A3R1 from human samples.PA1-27670 may be successfully used in ELISA and Western blot applications. By Western blot, this antibody has only been tested on placenta protein extract.The PA1-27505 immunogen is a synthetic peptide corresponding to human Ezrin-radixin-moesin.
Specifications

Specifications

SLC9A3R1
Polyclonal
PBS with no preservative
O14745
EBP50, NHERF, NHERF1, NPHLOP2
Chicken
IgY
50μg
Store at -20°C
Primary
9368
ELISA, Western Blot
Unconjugated
SLC9A3R1
Purified
SLC9A3R1
Synthetic peptide corresponding to human Ezrin-radixin-moesin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit