Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC9A9 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525327

Catalog No. PIPA525327

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a sodium hydrogen exchanger that is a member of the solute carrier 9 protein family. The encoded protein localizes the to the late recycling endosomes and may play an important role in maintaining cation homeostasis. Defects in this gene are associated with attention-deficit/hyperactivity disorder.
Specifications

Specifications

SLC9A9
Polyclonal
PBS with 0.09% sodium azide
Q8IVB4, Q8BZ00
AUTS16, NHE9, Na(+)/H(+) exchanger 9, putative protein product of Nbla00118, sodium/hydrogen exchanger 9, sodium/proton exchanger NHE9, solute carrier family 9 (sodium/hydrogen exchanger)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
285195, 331004
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SLC9A9
Liquid
SLC9A9
KLH conjugated synthetic peptide between 551-579 amino acids from the C-terminal region of human SLC9A9
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit