Learn More
SLCO4C1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human SLCO4C1 peptide using ARM Technology.
Supplier: Abnova Corporation H00353189K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
SLCO4C1 belongs to the organic anion transporter (OATP) family. OATPs are involved in the membrane transport of bile acids, conjugated steroids, thyroid hormone, eicosanoids, peptides, and numerous drugs in many tissues (Mikkaichi et al., 2004 [PubMed 14993604]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
SLCO4C1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
SLCO4C1 | |
SLCO4C1 | |
A synthetic peptide of human SLCO4C1 | |
RUO | |
353189 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
solute carrier organic anion transporter family, member 4C1 | |
SLCO4C1 | |
OATP-H, OATP-M1, OATP4C1, OATPX, PRO2176, SLC21A20 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.