Learn More
SLP-76 Mouse anti-Human, Mouse, Rat, Clone: AS55, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051426
Catalog No. 05-142-6MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect SLP-76 using this Anti-SLP-76 Antibody, clone AS55 validated for use in western blotting and immunoprecipitation.
Specifications
Specifications
SLP-76
AS55
Purified mouse monoclonal IgG2a 0.2mM sterile filtered solution in phosphate buffered saline with 0.1% sodium azide.
Purified
Murine
IgG2a
100μg
Primary
Human, Murine, Rat
Immunoprecipitation, Western Blot
Unconjugated
NM_005565.3
LCP2, SLP-76, SLP76
Monoclonal antibody from BALB/c-derived hybridoma
Protein G purfied
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal