Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLU7 Mouse anti-Human, Clone: P759, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA518304

Catalog No. PIMA518304


Add to cart

Description

Description

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Participates in the second catalytic step of pre-mRNA splicing, when the free hydroxyl group of exon I attacks the 3 ft.-splice site to generate spliced mRNA and the excised lariat intron. Required for holding exon 1 properly in the spliceosome and for correct AG identification when more than one possible AG exists in 3 ft.-splicing site region.
Specifications

Specifications

SLU7
Monoclonal
1 mg/mL
PBS with no preservative
O95391
hSlu7, SLU7 splicing factor homolog S. cerevisiae, 9G8
Mouse
IgG1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10569
Western Blot
P759
Unconjugated
SLU7
Liquid
SLU7
Recombit fragment corresponding to a region within amino acids 1 and 282 of SLU7
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit