Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAGLN Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527463

Catalog No. PIPA527463


Add to cart

Description

Description

PA5-27463 targets SM22 alpha in IF and WB applications and shows reactivity with Human, mouse and rat samples. The PA5-27463 immunogen is recombit fragment corresponding to a region within amino acids 1 and 201 of Human SM22 alpha. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a transformation and shape-change sensitive actin cross-linking/gelling protein found in fibroblasts and smooth muscle. Its expression is down-regulated in many cell lines, and this down-regulation may be an early and sensitive marker for the onset of transformation. A functional role of this protein is unclear. Two transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

TAGLN
Polyclonal
Unconjugated
TAGLN
Liquid
TAGLN
Recombit fragment corresponding to a region within amino acids 1 and 201 of Human SM22 alpha
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
0.96 mg/mL
PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Q01995, P37804, P31232
SM22, SMCC, TAGLN1, WS3-10, 22 kDa actin-binding protein, SM22-alpha, smooth muscle protein 22-alpha, transgelin variant 2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
21345, 25123, 6876
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit