Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smac/DIABLO Mouse, Unlabeled, Clone: 56, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 612246

Catalog No. BDB612246


Add to cart

Description

Description

Apoptosis is a genetically programmed, selective process of cell death that occurs during normal cell differentiation and development of multicellular organisms. Inhibitors of apoptosis proteins (IAPs), hILP/XIAP, c-IAP1, c-IAP2, and survivin, suppress apoptosis by preventing procaspase activation and inhibiting mature caspase activity. Smac/DIABLO (second mitochondria-derived activator of caspases) eliminates the inhibitory effect of many IAPs, and has been shown to interact with hILP, c-IAP1, c-IAP2, and survivin. The interaction with hILP occurs through Smac/DIABLO binding to the second and third BIR domains of hILP. Smac/DIABLO protein contains an N-terminal mitochondria targeting sequence (MTS) and a procaspase-3 activation domain. The N-terminal MTS is cleaved after import into the mitochondria, and the activity of the mature Smac/DIABLO is dependent on homodimerization and release from the mitochondria. Overexpression of Smac/DIABLO in HeLa cells increases the sensitivity to UV-mediated apoptosis. Thus, Smac/DIABLO acts as an apoptotic activator through its binding to IAPs and activation of caspases.

Western Blotting

Specifications

Specifications

Smac/DIABLO
56
Unconjugated
Affinity Purified
Human Smac/DIABLO aa. 125-239
50μg
Cell Biology
Primary
Canine, Human, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2b
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit