Learn More
anti-SMAD1/5/8/9, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Manufacturer:  Novus Biologicals NB10056443
 View more versions of this product
Catalog No. NB10056443
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
SMAD1/5/8/9 Polyclonal antibody specifically detects SMAD1/5/8/9 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
Specifications
SMAD1/5/8/9
0.5 mg/mL
Western Blot 1 - 3 ug/mL
Q15797
BSP1, BSP-1, hSMAD1, JV4-1SMAD, mothers against DPP homolog 1 (Drosophila), MAD homolog 1, MADH1JV41, MADR1MAD, mothers against decapentaplegic homolog 1 (Drosophila), Mad-related protein 1, mothers against decapentaplegic homolog 1, Mothers against DPP homolog 1, SMAD family member 1SMAD 1, SMAD, mothers against DPP homolog 1, Smad1, TGF-beta signaling protein 1, transforming growth factor-beta signaling protein 1
Rabbit
IgG
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
In A549, a 50 kDa band is observed. The two synthetic peptide immunogens (LGWKQGDEEEKWAEK) and (SNVNRNSTIENTRRHI) used to make this antibody are 100% conserved between SMAD1/5/8/9. Therefore, the antibody recognizes SMADs 1, 5, 8, and 9. The antibody may cross-react with SMAD2 or SMAD3. The actual SMADs detected with this antibody may vary according to cell or tissue-type specific expression.
Western Blot
Unconjugated
PBS, 0.05% BSA with 0.05% Sodium Azide
Purified
SMAD1
This antibody was developed against two synthetic peptides corresponding to amino acids 18-32 (LGWKQGDEEEKWAEK) and amino acids 315-330 (SNVNRNSTIENTRRHI) of human SMAD1. GenBank Accession No. AAP36050.1.
52 kDa
0.1 mg
Cancer, Diabetes Research
Primary
4086
Human, Mouse, Rat