Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAD1/SMAD5/SMAD9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141238

Catalog No. 01-675-592


Add to cart

Description

Description

This antibody is predicted to react with chicken, chimpanzee, invertebrates, most mammals, porcine, xenopus and zebrafish samples

SMADs are members of the MAD-related family of molecules. MAD-related proteins are a  family of intracellular proteins that are essential components in the signaling pathways of the serine/threonine kinase receptors of the transforming growth factor beta superfamily (1).  SMADs can be divided into receptor-regulated SMADs (R-SMADs: SMAD1, SMAD2, SMAD5, SMAD8 and SMAD9), common-mediator SMAD (co-SMAD: SMAD4), and inhibitory SMADs (I-SMADs: SMAD6 and SMAD7).  SMAD1, SMAD5, SMAD8 and SMAD9 have high degrees of homology and antibodies are available that recognize sequences common to all of them.  SMAD8 and SMAD9 are typically used as alternate names for one another in the literature. 
Specifications

Specifications

SMAD1/SMAD5/SMAD9
Polyclonal
Unconjugated
SMAD1
Liquid
SMAD1, SMAD5, SMAD9
Synthetic peptides corresponding to residues L(18) G W K Q G D E E E K W A E K(32) and amino acids S(315) N V N R N S T I E N T R R H I(330) of human SMaD1.
Protein G
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
O15198, Q15797, Q99717, P70340, P97454, Q9JIW5, O54835, P97588, Q9R1V3
Smad1, SMAD5, SMAD9, BSP1, JV4-1, JV41, MADH1, MADR1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
17125, 17129, 25671, 4086, 4090, 4093, 55994, 59328, 85435
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit