Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMAD1 Mouse anti-Human, Mouse, Clone: AS22, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051459
Description
Anti-SMAD1 Antibody, clone AS22 detects level of SMAD1 and has been published and validated for use in western blotting and immunofluoresence.Specifications
SMAD1 | |
Monoclonal | |
Unconjugated | |
NP_001003688 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunofluorescence, Western Blot | |
AS22 | |
Purified mouse monoclonal in 0.1M Tris-Glycine (pH7.4), 150mM NaCl with 0.05% NaN3. | |
BSP-1 , BSP1, CMT2D , GARS, HMN5, JV4-1, JV41, MADH1, MADR1 , hSMAD1 | |
GST-fusion protein corresponding to amino acids 1-110 of human SMAD1. | |
200 μg | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only