Learn More
Smad2/3 Mouse, Unlabeled, Clone: 18, BD
Mouse Monoclonal Antibody
$255.00 - $505.00
Specifications
Antigen | Smad2/3 |
---|---|
Clone | 18 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The transforming growth factor β (TGFβ)/activin/BMP family of growth factors plays a diverse and important role in growth, development, and differentiation. These growth factors act through their binding to heteromeric plasma membrane receptor protein kinases which, upon ligand binding, become activated and trigger an intracellular signaling cascade. Specifically, receptor activation induces the translocation of a set of conserved proteins named Smads (Sma- and Mad-related proteins) to the nucleus, resulting in gene activation. Smad2 is a ubiquitously expressed protein of 58kDa that is phosphorylated and translocated to the nucleus in response to TGFβ, but not BMP. The overall response to TGFβ is growth inhibition. The Smad2 gene is located in chromosome 18q21.1 which is often absent in several human cancers. Furthermore, some missense mutations on the Smad2 gene were identified in colorectal carcinomas, suggesting Smad2 may function as a tumor suppressor in normal cells.
Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
Smad2/3 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
18 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Mouse Smad2 aa. 142-263 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.