Learn More
Smad2/3 Rabbit anti-Human, Mouse, Rat, Clone: C4T, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier:  MilliporeSigma 04914
Catalog No. 04-914-MI
Add to Cart
Description
Description
Use Anti-Smad2/3 Antibody, clone C4T (Rabbit Monoclonal Antibody) validated in western blotting to detect Smad2/3 also known as Deletion target in pancreatic carcinoma 4, mothers against decapentaplegic homolog 4.
Specifications
Specifications
Smad2/3
Monoclonal
Unconjugated
NP_001003652.1, NP_005893.1
Fusion protein corresponding to amino acids 186-273 of human Smad2.
Primary
Human, Mouse, Rat
Purified
Western Blot
C4T
Cultured supernantant containing 0.05% sodium azide.
Rabbit
100 μL
DPC4, JIP, MADH4, OTTHUMP00000163548, hSMAD4
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Recommendations
Recommendations