Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Smad2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NB10056462SS
Description
Smad2 Polyclonal specifically detects Smad2 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Smad2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2 | |
Rabbit | |
Protein G purified | |
RUO | |
Primary | |
The SMAD2 amino acid sequence used as immunogen is 100% homologous in human (isoform 1: 467 amino acids; isoform 2: 437 amino acids), mouse, rat, horse, cow, chicken and Xenopus. | |
Store at 4C. Do not freeze. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
NP_005892 | |
SMAD2 | |
Amino acids 234-249 (DQQLNQSMDTGSPAEL) of human SMAD2 protein was used as the immunogen. | |
0.025 mg | |
Cancer, Cell Cycle and Replication, Metastasis, Wnt Signaling Pathway | |
4087 | |
Human, Mouse, Rat, Chicken | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction