Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAGP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525195

Catalog No. PIPA525195

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

SMAGP may play a role in epithelial cell-cell contacts. SMAGP may play a role in tumor invasiveness and metastasis formation.
Specifications

Specifications

SMAGP
Polyclonal
PBS with 0.09% sodium azide
Q0VAQ4
hSMAGP, small cell transmembrane and glycosylated protein, small trans-membrane and glycosylated protein, small transmembrane and glycosylated protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57228
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SMAGP
Liquid
SMAGP
KLH conjugated synthetic peptide between 46-75 amino acids from the central region of human SMAGP
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit