Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLTF, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Supplier:  Thermo Scientific PA131579

Catalog No. PIPA131579

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

The HLTF (Helicase-like Transcription Factor; SMARCA 3) gene is a member of the family of genes that remove proteins that coil around DNA. HLTF proteins aid the stabilization and regulation of the production of proteins in the cell. Inactivation of the HT

PA1-31579 detects SMARCA3 from human samples.PA1-31579 has been successfully used in Western blot applications.The PA1-31579 immunogen is: Peptide with sequence KQAKINEIRTLIDL, from C Terminus of the protein sequence according to NP_003062; NP_620636
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SMARCA3
Polyclonal
Unconjugated
Q14527, Q6PCN7
HLTF
Peptide with sequence KQAKINEIRTLIDL, from C Terminus of the protein sequence according to NP_003062; NP_620636
50 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
0.5mg/mL
HLTF
HIP116, HIP116A, HLTF1, RNF80, SMARCA3, SNF2L3, ZBU1
Goat
IgG fraction
RUO
6596
Store at -20°C, Avoid Freeze/Thaw Cycles
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit