Learn More
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SMARCC1.
Supplier: Abnova Corporation H00006599A01
Description
The protein encoded by this gene is a member of the SWI/SNF family of proteins, whose members display helicase and ATPase activities and which are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI and contains a predicted leucine zipper motif typical of many transcription factors. [provided by RefSeq
Sequence: ESRKKSGKKGQASLYGKRRSQKEEDEQEDLTKDMEDPTPVPNIEEVVLPKNVNLKKDSENTPVKGGTVADLDEQDEETVTAGGKEDEDPAKGDQSRSVDLSpecifications
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SMARCC1. | |
SMARCC1 | |
BAF155/CRACC1/Rsc8/SRG3/SWI3 | |
Mouse | |
50 μL | |
Transcription Regulation | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_003074 | |
SMARCC1 | |
SMARCC1 (NP_003065, 338 a.a. ∼ 437 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
6599 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.