Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMCR7L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525483

Catalog No. PIPA525483


Add to cart

Description

Description

SMCR7L belongs to the SMCR7 family. The specific function of this protein remains unknown.
Specifications

Specifications

SMCR7L
Polyclonal
PBS with 0.09% sodium azide
L0R8F8, Q8BGV8
HSU79252, MID51, MIEF1, dJ1104E15.3, mitochondrial dynamic protein MID51, mitochondrial dynamic protein of 51 kDa, mitochondrial elongation factor 1, smith-Magenis syndrome chromosomal region candidate gene 7 protein-like
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
239555, 54471
Western Blot
Unconjugated
MIEF1
Liquid
MIEF1
KLH conjugated synthetic peptide between 105-132 amino acids from the N-terminal region of human SMCR7L
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit