Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMN Mouse anti-Human, Mouse, Xenopus, Clone: 62E7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 041466
Description
Use Anti-SMN Antibody, clone 62E7 (Mouse Monoclonal Antibody) validated in western blot, immunocytochemistry, immunoprecipitation to detect SMN also known as Spinal muscular atrophy (Werdnig-Hoffmann disease Kugelberg-Welander disease).Specifications
SMN | |
Monoclonal | |
Unconjugated | |
NP_000335.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
62E7 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
SMN1, BCD541, Gemin-1, SMA, SMA1, SMA2, SMA3, SMA4, SMA@, SMN, SMNC, SMNT | |
His-tagged recombinant protein corresponding to human SMN. | |
100 μg | |
Human, Mouse, Xenopus species | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only