Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SMN2 Ab-4, MaxPab™ Polyclonal, Abnova™

Mouse Polyclonal

Manufacturer:  Abnova Corporation H00006607B01

Catalog No. 89-002-502


Add to cart

Description

Description

Host Species: Mouse
Cross Reactivity: Human
Immunogen: SMN2 (NP_075013.1, 1 a.a. ~282 a.a) full-length human protein.

Detection Antibody, Western Blotting

Specifications

Specifications

SMN2
Unconjugated
No additive
NM_022875
BCD541|C-BCD541|FLJ76644|MGC20996|MGC5208|SMNC
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human SMN2 protein.
SMN2
Liquid
SMN2
SMN2 (NP_075013.1, 1 a.a. ~ 282 a.a) full-length human protein.
RUO
Primary
6607
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit