Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMNDC1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526379

Catalog No. PIPA526379

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

This protein is a paralog of SMN1 gene, which encodes the survival motor neuron protein, mutations in which are cause of autosomal recessive proximal spinal muscular atrophy. The protein encoded by this gene is a nuclear protein that has been identified as a constituent of the spliceosome complex. This protein is differentially expressed, with abundant levels in skeletal muscle, and may share similar cellular function as the SMN1 gene.
Specifications

Specifications

SMNDC1
Polyclonal
PBS with 0.09% sodium azide
O75940, Q8BGT7
SMNR, SPF30, 30 kDa splicing factor SMNrp, SMN-related protein, splicing factor 30, survival of motor neuron-related, survival motor neuron domain-containing protein 1, survival of motor neuron-related-splicing factor 30
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10285, 76479
Flow Cytometry, Western Blot
Unconjugated
SMNDC1
Liquid
SMNDC1
KLH conjugated synthetic peptide between 11-41 amino acids from the N-terminal region of human SMNDC1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit