Learn More
MilliporeSigma™ anti-SMO Clone: 1D9, Available on GSA/VA Contract for Federal Government customers only.
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma™ ST1718100UG
Catalog No. ST1718100UG
Add to cart
Description
Description
Specifically detects SMO Clone: 1D9 in Human samples, and it is validated for Immuno Blotting
SMO is a G protein-coupled receptor that probably associates with the protein, Patched (PTCH), to transduce hedgehog signalling. Binding of sonic hedgehog (SHH) to its receptor, Patched, is thought to inhibit the interaction between SMO and Patched.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
SMO
1D9
In PBS, pH 7.2.
Murine
IgG2a
RUO
Primary
Recognizes the ∽75kDa SMO protein in A431 cells.
Immuno Blotting, Immunoblot
Unconjugated
Purified
Full-length, recombinant, human SMO (aa 653-787) expressed as a GST fusion protein
100μg
−20°C
Monoclonal
Human