Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Smooth Muscle Actin, clone: 1A4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA137027

Catalog No. 01-671-636

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody stains smooth muscle cells in vessel walls, gut wall, and myometrium. Myoepithelial cells in breast and salivary gland are also stained. It reacts with tumors arising from smooth muscles and myoepithelial cells. Actin from fibroblasts (beta- and gamma-cytoplasmic), striated muscle (alpha-sarcomeric), and myocardium (alpha-myocardial) are not stained.

This antibody is predicted to react with mouse, rat, rabbit, baboon, and bovine based on sequence homology.A suggested positive control is small intestine (smooth muscle) or leiomyoma.
Specifications

Specifications

Smooth Muscle Actin
Monoclonal
Unconjugated
ACTA2
Purified
ACTA2
Synthetic peptide from N-terminus of human alpha smooth muscle isoform
Protein A
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin)
1A4
PBS with 1% BSA and <0.1% sodium azide; pH 7.6
P62736
AAT6, ACTSA, alpha-sma
Murine
IgG1, κ
1mL
Store at 4°C
Primary
59
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit