Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMRT Mouse anti-Human, Mouse, Rat, Clone: 1SM-1E7, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 041551
Description
Use Anti-SMRT Antibody, clone 1SM-1E7 (Mouse Monoclonal Antibody) validated in western blotting to detect SMRT also known as CTG repeat protein 26, Silencing mediator of retinoic acid and thyroid hormone receptor.Specifications
SMRT | |
Monoclonal | |
Unconjugated | |
NP_001070729.1 | |
Mouse | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG2a κ |
Western Blot | |
1SM-1E7 | |
Unpurified mouse monoclonal IgG2aκ ascites with 0.05% sodium azide. | |
NCOR2, CTG26, N-CoR2, SMAP270, SMRT, SMRTE, SMRTE-tau, TNRC14, TRAC, TRAC-1, TRAC1 | |
Recombinant protein corresponding to human SMRT. | |
100 μL | |
Human, Mouse, Rat | |
Ascites |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only