Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMRT Mouse anti-Human, Unlabeled, Clone: 44, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611386

 View more versions of this product

Catalog No. BDB611386


Add to cart

Description

Description

Steroid nuclear hormone receptors regulate physiological homeostasis through repression and activation of gene transcription. In the absence of hormone, DNA-bound steroid receptors recruit corepressor, such as silencing mediator of retinoic acid and thyroid hormone receptor (SMRT) and nuclear receptorcorepressor (N-CoR), which bind to the free ligand binding domain of the thyroid hormone receptor, retinoic acid receptor, and other nuclear receptors. SMRT and N-CoR form a large protein complex with histone deacetylase I (HDAC1) and Sin3A, which deacetylates histones to alter chromatin structure in a manner that inhibits transcription. In addition, SMRT and N-CoR proteins interact with the transcription factor CBF/RBP-Jk to regulate Notch signaling pathways. SMRT contains four domains related to repressor activity (RD1, RD2, SRD1, and SRD2), two receptor interaction domains (RID1 and RID2), and a SANT region that is found in yeast chromatin remodeling factor SWI3. The ubiquitous expression pattern and corepressor activity of SMRT indicates that this protein is important for hormonal control of gene transcription in many tissues.

Immunofluorescence, Western Blotting

Specifications

Specifications

SMRT
44
Unconjugated
Affinity Purified
Human SMRT aa. 1366-1473
50μg
Cell Biology
Primary
Human
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.