Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUMO-3 Rabbit anti-Equine, Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523110

Catalog No. PIPA523110


Add to cart

Description

Description

SUMO3 (SMT3) is a Ubiquitin-like protein found in Saccharomyces cerevisiae similar to the mammalian Ubiquitin-like SUMO1, which is involved in posttranslational protein modification. SUMO3 is functionally active in regulation of nucleocytoplasmic transport and suggest a role in cell-cycle regulation. Knockdown of SUMO3 in Drosophila in the prothoracic gland, which controls key developmental processes through the synthesis and release of ecdysteroids, caused a 4-fold prolongation of larval life and completely blocked the transition from larval to pupal stages.
Specifications

Specifications

SUMO-3
Polyclonal
Unconjugated
SUMO3
Liquid
SUMO3
Synthetic peptide (71 CDTPAQLEMEDE 81) of SMT3/SUMO 3 protein.
Protein A
RUO
Antibody
Polyclonal
Equine, Human, Mouse, Non-human primate
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P55854, Q9Z172
SMT3A, SMT3H1, SUMO-3, Smt3B, SMT3 homolog 1, SMT3 suppressor of mif two 3 homolog 1, SUMO-2, small ubiquitin-related modifier 3, ubiquitin-like protein SMT3A, ubiquitin-like protein SMT3B
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100630015, 20610, 6612
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit