Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SMYD3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220430
Description
SMYD3 Polyclonal antibody specifically detects SMYD3 in Human, Mouse samples. It is validated for Western Blot, ChIP assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation.Specifications
SMYD3 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 | |
SMYD3 | |
Recombinant protein encompassing a sequence within the center region of human SMYD3. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
64754 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, ChIP assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
0.79mg/mL | |
Western Blot 1:500-1:3000, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500 | |
bA74P14.1, bA74P14.1 (novel protein), EC 2.1.1, EC 2.1.1.43, FLJ21080, KMT3E, MGC104324, MYND domain containing 1, SET and MYND domain containing 3, SET and MYND domain-containing protein 3, Zinc finger MYND domain-containing protein 1, zinc finger protein, subfamily 3A (MYND domain containing), 1, ZMYND1, ZNFN3A1 | |
Rabbit | |
49 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only