Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAIL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523482

Catalog No. PIPA523482


Add to cart

Description

Description

Zinc finger protein SNAI1 (SNAIL)is a zinc finger transcriptional repressor which down regulates the expression of the ectodermal gene within the mesoderm and functions to form the mesoderm and neural crest. It is reported that SNAI1 acts as an important factor of tumor invasion as evidenced by its role in E-cadherin down-regulation and induction of epithelial-mesenchymal transition (EMT). In human breast cancer, the expression of SNAI1 and/or the homologous SNAI2 (Slug) has been associated with E-cadherin repression, local or distant metastasis, tumor recurrence, or poor prognosis in different tumor series. It is also reported that Snail 1 protein in the stromamay be anew putative prognosis marker for colon tumors.
Specifications

Specifications

SNAIL
Polyclonal
Unconjugated
SNAI1
Liquid
SNAI1
Synthetic peptide (3 RSFLVRKPSDPNRKPNYS 20) of SNAI1/Snail protein.
Protein A
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.0 mg/mL
PBS with 0.05% sodium azide
O95863
SLUGH2, SNA, SNAH, SNAIL, SNAIL1, dJ710H13.1, protein sna, protein snail homolog 1, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, zinc finger protein SNAI1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6615
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit