Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAI2/Slug Mouse, PE, Clone: S43-1259, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564615

Catalog No. BDB564615


Add to cart

Description

Description

The S43-1259 monoclonal antibody specifically recognizes SNAI2, also named Slug, which is a zinc finger transcriptional repressor first recognized for its roles in embryogenesis. SNAI2/Slug and other members of the snail family of transcription factors inhibit the expression of E-cadherin, which plays important roles in cell adhesion and in the maintenance of tissue structure. SNAI2/Slug activation and E-cadherin inhibition induces epithelial-mesenchymal transition (EMT) at several stages of embryonic development and also contributes to the invasiveness of malignancies. Aberrant expression of SNAI2/Slug has also been linked to cancer stem cell formation, cell cycle regulation and apoptosis. SNAI2/Slug is a downstream effector of various signaling pathways, including Akt, Wnt and SCF/ckit signaling.

Specifications

Specifications

SNAI2/Slug
S43-1259
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Snai2; SLUG; Slugh; SLUGH
Affinity Purified
Human SNAI2 Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit