Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNAP29 Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen OSS00051G

Catalog No. PIOSS00051G


Add to cart

Description

Description

Reconstitute with 500 µL of distilled water.

Specifications

Specifications

SNAP29
Polyclonal
Unconjugated
Snap29
Lyophilized
SNAP29
A synthetic peptide from rat Synaptosomal-associated protein 29
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Mouse, Rat
Immunohistochemistry, Western Blot
1 mg/mL
Antibody with no preservative
Q9ERB0, Q9Z2P6
SNAP-29, Vesicle-membrane fusion protein, SNAP29, Soluble 29 kDa NSF attachment protein, Golgi SNARE of 32 kDa, Gs32
Rabbit
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
116500, 67474
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit