Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

SNAP alpha Mouse anti-Bovine, Human, Rat, Clone: 4E4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112452

Catalog No. 01-674-319



Description

Description

A suggested positive control for this product is rat brain lysate.

SNAPs (solubleNSFattachment proteins), acting in concert with SNAREs ( S N A P r e c eptors) and the N - ethylmaleimide- s e nsitive f u sion protein (NSF) are required for the fusion of transport vesicles to their target membranes in synaptic transmission, intra-Golgi transport, endosome-toendosome fusion and transcytotic vesicles-to-plasma membrane transport. Vesicle-to-target membrane docking (initial contact) occurs when the vesicle SNARE binds to its cognate target membrane SNARE. a-SNAP (or b-SNAP in brain) then binds to this docking complex and mediates the binding of NSF and thus the formation of a 20S fusion particle. It is thought that, once NSF is bound, ATP hydrolysis by NSF initiates the fusion process.
Specifications

Specifications

SNAP alpha
Monoclonal
Unconjugated
NAPA
Liquid
NAPA
Human a-SNAP protein.
Purified
RUO
Antibody
Monoclonal
Bovine, Human, Rat
Immunoprecipitation, Western Blot
4E4
PBS with 50% glycerol and no preservative; pH 7.5
P81125, P54920, P54921
Synaptosomal-Associated Protein alpha
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
140673, 518407, 8775
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit