Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SNF1LK2/SIK2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP142690

Catalog No. NBP142690


Add to cart

Description

Description

SNF1LK2/SIK2 Polyclonal antibody specifically detects SNF1LK2/SIK2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

SNF1LK2/SIK2
1.0 mg/mL
Western Blot, Immunoprecipitation 2-5 ug/mg lysate
Affinity Purified
SIK2
The immunogen for this product maps to a region between residue 600 and 650 of human salt-inducible kinase 2 using the numbering given in entry NP_056006.1 (GeneID 23235).
Immunogen affinity purified
RUO
Store at 4C. Do not freeze.
Polyclonal
Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon.
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
DKFZp434K1115, EC 2.7.11, EC 2.7.11.1, KIAA0781SIK-2, LOH11CR1I, QIKSNF1LK2, Qin-induced kinase, salt-inducible kinase 2, Salt-inducible protein kinase 2, salt-inducible serine/threonine kinase 2, serine/threonine-protein kinase SIK2, Serine/threonine-protein kinase SNF1-like kinase 2, SNF1-like kinase 2
Rabbit
IgG
100 ul
Protein Kinase
Primary
23235
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only