Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNRPC Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512458

Catalog No. PIPA512458

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, mouse and non-human primate based on sequence homology.

SNRPC is associated with snRNP U1.
Specifications

Specifications

SNRPC
Polyclonal
PBS with 0.09% sodium azide
P09234
U1 small nuclear ribonucleoprotein C; U1 snRNP protein C; U1C protein; U1-C; SNRPC
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6631
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
SNRPC
Liquid
SNRPC
KLH conjugated synthetic peptide between 133-159 amino acids from the C-terminal region of human SNRPC
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit