Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-SNTB2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133009

Catalog No. NBP133009


Add to cart

Description

Description

SNTB2 Polyclonal antibody specifically detects SNTB2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

SNTB2
0.84mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
SNTB2
Recombinant protein encompassing a sequence within the center region of human SNTB2. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
6645
Human
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
D16S2531E, EST25263, SNT2B2dystrophin-associated protein A1, 59kD, basic component 2, SNT3beta-2-syntrophin, SNTLsyntrophin-3, syntrophin, beta 2 (dystrophin-associated protein A1, 59kD, basic component 2), syntrophin, beta 2 (dystrophin-associated protein A1, 59kDa, basic component 2), Syntrophin-3,59 kDa dystrophin-associated protein A1 basic component 2, Syntrophin-like
Rabbit
IgG
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only