Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA529543
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (87%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SNX1 Polyclonal antibody specifically detects SNX1 in Human samples. It is validated for Western Blot.Specifications
SNX1 | |
Polyclonal | |
Unconjugated | |
SNX1 | |
HsT17379; MGC8664; Snx1; SNX1A; sorting nexin 1; sorting nexin 1A; sorting nexin-1; VPS5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6642 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q13596 | |
SNX1 | |
Recombinant fragment corresponding to a region within amino acids 189 and 504 of Human SNX1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction