Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SNX2, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127870

Catalog No. PA127870

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in several stages of intracellular trafficking.

PA1-27870 detects SNX2 from human, mouse, and rat samples.PA1-27870 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Sorting nexin-2.
Specifications

Specifications

SNX2
Polyclonal
PBS with no preservative
O60749, Q9CWK8
MGC5204, TRG-9
Chicken
IgY
50μg
Store at -20°C
Primary
6643
Western Blot
Unconjugated
SNX2
Purified
SNX2
Synthetic peptide corresponding to human Sorting nexin-2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit