Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX27 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
$204.00 - $482.50
Specifications
Antigen | SNX27 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1 ug/ml, Simple Western 10 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation reported in scientific literature (PMID 32413996), Immunohistochemistry-Paraffin 1:100 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP145283SS
|
Novus Biologicals
NBP145283SS |
0.025 mL |
Each for $204.00
|
|
|||||
NBP145283
|
Novus Biologicals
NBP145283 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
SNX27 Polyclonal specifically detects SNX27 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
SNX27 | |
Western Blot 1 ug/ml, Simple Western 10 ug/ml, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation reported in scientific literature (PMID 32413996), Immunohistochemistry-Paraffin 1:100 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS and 30% Glycerol with 0.1% Sodium Azide | |
KIAA0488MGC126873, methamphetamine-responsive transcript 1, MGC126871, MGC20471, MRT1, MY014, sorting nexin family member 27, sorting nexin-27 | |
SNX27 | |
IgG | |
Affinity Purified | |
Human and mouse. Predicted to react with rat, bovine and Zebrafish based on 100% sequence homology. |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Q96L92, Q3UHD6 | |
81609 | |
Synthetic peptide made to an internal portion of human SNX27 (within residues 450-500). [Swiss-Prot# Q96L92] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
65 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title