Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-SOCS-3, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$175.82 - $378.00
Specifications
Antigen | SOCS-3 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Immunohistochemistry-Paraffin 5ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227116SS
|
Novus Biologicals
NBP227116SS |
0.025 mg |
Each for $175.82
|
|
NBP227116
|
Novus Biologicals
NBP227116 |
0.1 mg |
Each for $378.00
|
|
Description
SOCS-3 Polyclonal specifically detects SOCS-3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SOCS-3 | |
Immunohistochemistry-Paraffin 5ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3 | |
SOCS3 | |
IgG | |
Protein A purified | |
The SOCS3 amino acid sequence used as immunogen for this antibody is 100% homologous in human, mouse, rat, cow, guinea pig, dog, Chinese hamster, horse, mole rat, elephant, rhesus monkey, sheep, pig, and pole-cat, 94% in panda, Carolina anole, new world monkey, chicken, wild turkey, opossum, and zebra finch, 89% in platypus, 72% in channel catfish, and 67% homologous in frog. |
0.5 mg/mL | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
9021 | |
A portion of amino acids 170-220 from human SOCS3 was used as the immunogen for this antibody. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title