Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOCS5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521600

Catalog No. PIPA521600


Add to cart

Description

Description

PA5-21600 targets SOCS5 in IHC (P) and WB applications and shows reactivity with Human and Mouse samples. The PA5-21600 immunogen is a synthetic peptide corresponding to a region within amino acids 1 and 42 of SOCS5. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

SOCS5
Polyclonal
Unconjugated
SOCS5
Liquid
SOCS5
Synthetic peptide contain a sequence corresponding to a region within amino acids 1 and 42 of SOCS5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
O75159, O54928
CIS6, CISH6, Cish5, SOCS-5, CIS-6, cytokine-inducible SH2 protein 6, cytokine-inducible SH2-containing protein 5
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
56468, 9655
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit