Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD1 Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523245

Catalog No. PIPA523245


Add to cart

Description

Description

SOD1 (superoxide dismutase) binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. This isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene.
Specifications

Specifications

SOD1
Polyclonal
Unconjugated
SOD1
Liquid
SOD1
superoxide dismutase enzyme from bovine erythrocytes
Protein A
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
1.0 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P00442, P00441, P08228, P07632
ALS, ALS1, IPOA, SOD, hSod1, homodimer, Cu/Zn superoxide dismutase, SOD, soluble, indophenoloxidase A, superoxide dismutase [Cu-Zn], superoxide dismutase, cystolic
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
20655, 24786, 281495, 6647
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit