Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD2, Polyclonal, Invitrogen™

Catalog No. PIPA125346
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA125346 50 μg
1 options

Catalog No. PIPA125346

Supplier: Thermo Scientific PA125346

Goat Polyclonal Antibody

manganese superoxide dismutase; intramitochondrial free radical scavenging enzyme.

PA1-25346 detects MnSOD from rat, mouse, and human samples. PA1-25346 is expected to cross react with human and mouse due to sequence homology.PA1-25346 has been successfully used in Western blot procedures.The PA1-25346 immunogen is a synthetic peptide corresponding to a portion of human MnSOD encoded within exon 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOD2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene SOD2
Gene Accession No. P04179, P09671, P07895
Gene Alias Manganese superoxide dismutase
Gene Symbols SOD2
Host Species Goat
Immunogen Synthetic peptide corresponding to a portion of human MnSOD encoded within exon 5.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6648
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.