Learn More
Sodium/Calcium Exchanger, clone: 6H2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA112751
Catalog No. 01-674-052
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Ca2+ plays a critical role in intracellular signaling and thus regulation of calcium balance is critical for the maintenance of this signaling function. The plasma membrane Na+/Ca2+ (NaCX) exchangers mediate Na+-dependent Ca2+ efflux in a wide variety of cell types. NaCX can move Ca2+ either into or out of cells, depending on the net Na+, Ca2+, and K+ gradient across the membrane. In mammals, at least 5 distinct genes code for the exchangers: Three NaCX (NaCX-1, NaCX-2, and NaCX-3), and two in the NaCKX family (NaCKX-1 and NaCKX-2). NaCX-1 is most prominently expressed in the heart where it plays a major role in excitation-contraction coupling (Matsuoka et al., 1993). NACX-1 may also be a target for protection of the heart and brain from ischemia and reperfusion injury (Hagihara et al., 2005; Bano et al., 2005; Choi, 2005).
Reconstitute with 50 ul of PBS.
Specifications
Specifications
Sodium/Calcium Exchanger
Monoclonal
Unconjugated
SLC8A1
Sodium/Calcium Exchanger 1
Murine
IgG2b
10 Miniblots
-20° C, Avoid Freeze/Thaw Cycles, Avoid Freeze/Thaw Cycles
Primary
100328570
Western Blot
6H2
Antibody with no preservative
Lyophilized
SLC8A1
Fusion protein from the N-terminus of rabbit renal Na+/Ca2+ exchanger.
Protein A/G
RUO
Antibody
Monoclonal
Rabbit