Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sodium Channel Nav1.7 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5390200UL
Description
Anti-Sodium Channel Nav1.7 Antibody, Pain detects level of Sodium Channel Nav1.7 and has been published and validated for use in western blotting.
Specifications
Sodium Channel Nav1.7 | |
Polyclonal | |
Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.025% sodium azide as a preservative. Reconstitute with 200μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 | |
Rabbit | |
Affinity Purified | |
Primary | |
Rat |
Western Blot | |
Unconjugated | |
SCN9A; NE-NA; NENA; ETHA; Nav1.7; hNE-Na; PN1 | |
Purified peptide from rat PN1 (amino acids 446-460) (Accession AAB50403) | |
200 μL | |
NM_002977.2 | |
Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction