Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A5 Rabbit anti-Human, Porcine, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA136151

Catalog No. 01-672-104


Add to cart

Description

Description

Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.

This gene encodes a member of the sodium glucose cotransporter family. The encoded protein is responsible for the uptake of iodine in tissues such as the thyroid and lactating breast tissue. The iodine taken up by the thyroid is incorporated into the metabolic regulators triiodothyronine (T3) and tetraiodothyronine (T4). Mutations in this gene are associated with thyroid dyshormonogenesis 1.
Specifications

Specifications

SLC5A5
Polyclonal
whole serum with 0.1% sodium azide
Q92911, Q63008
NIS, TDH1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Porcine, Rat
Immunohistochemistry (Frozen)
Unconjugated
SLC5A5
Lyophilized
SLC5A5
Synthetic peptide from rat NIS.
50 μL
4° C
Primary
114613, 399542, 6528
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit